Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iglon5
Entrez Gene ID
100754556
Gene Name
IgLON family member 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615616: 162106 - 167708 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653252.1 XP_007651442.1 3 - 4 337 69

View Full Product Details