Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc66
Entrez Gene ID
100756871
Gene Name
leucine rich repeat containing 66
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615260: 109709 - 128990 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652396.1 XP_007650586.1 4 - 5 999 97

View Full Product Details