Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dok7
Entrez Gene ID
100765890
Gene Name
docking protein 7
Gene Aliases
I79_002032
Chromosome Location
Chr.Un|NW_003613629: 334856 - 357622 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496603.1 XP_003496651.1 2 - 3 423 78

View Full Product Details