Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsen54
Entrez Gene ID
100755913
Gene Name
TSEN54 tRNA splicing endonuclease subunit
Gene Aliases
I79_000920
Chromosome Location
Chr.Un|NW_003613598: 89770 - 100228 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495697.2 XP_003495745.1 4 - 5 525 55

View Full Product Details