Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnk17
Entrez Gene ID
100770528
Gene Name
potassium channel, subfamily K, member 17
Gene Aliases
I79_007212
Chromosome Location
Chr.Un|NW_003613818: 810474 - 820557 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500897.1 XP_003500945.1 1 - 2 179 66

View Full Product Details