Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bnc1
Entrez Gene ID
100768562
Gene Name
basonuclin 1
Gene Aliases
I79_017795
Chromosome Location
Chr.Un|NW_003614736: 133287 - 239439 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509596.1 XP_003509644.1 3 - 4 308 102

View Full Product Details