Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sstr1
Entrez Gene ID
100766048
Gene Name
somatostatin receptor 1
Gene Aliases
I79_019446
Chromosome Location
Chr.Un|NW_003615006: 7607 - 24714 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511009.2 XP_003511057.1 2 - 3 364 94

View Full Product Details