Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmx2
Entrez Gene ID
100768285
Gene Name
H6 family homeobox 2
Gene Aliases
I79_018863
Chromosome Location
Chr.Un|NW_003614900: 272388 - 274828 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510538.2 XP_003510586.1 1 - 2 73 102

View Full Product Details