Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrit1
Entrez Gene ID
100753339
Gene Name
leucine-rich repeat, immunoglobulin-like and transmembrane domains 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614076: 449859 - 457795 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504559.1 XP_003504607.1 2 - 3 595 86

View Full Product Details