Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab40b
Entrez Gene ID
100752351
Gene Name
RAB40B, member RAS oncogene family
Gene Aliases
I79_018229
Chromosome Location
Chr.Un|NW_003614801: 451934 - 481155 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509989.2 XP_003510037.2 3 - 4 367 118

View Full Product Details