Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nsun3
Entrez Gene ID
100769254
Gene Name
NOP2/Sun domain family, member 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613645: 1996517 - 2046491 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007609741.1 XP_007607931.1 3 - 4 510 81
GenBank mRNA JI864047.1 - 3 - 4 516 81
JP042244.1 - 3 - 4 516 81

View Full Product Details