Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Parp16
Entrez Gene ID
100753348
Gene Name
poly (ADP-ribose) polymerase family, member 16
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614337: 423978 - 447956 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647711.1 XP_007645901.1 6 - 7 1100 83
XM_007647712.1 XP_007645902.1 6 - 7 1043 83
GenBank mRNA JI887785.1 - 3 - 4 514 83

View Full Product Details