Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Paip1
Entrez Gene ID
100764567
Gene Name
poly(A) binding protein interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614106: 906770 - 933550 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645443.1 XP_007643633.1 7 - 8 890 96
XM_007645444.1 XP_007643634.1 7 - 8 875 96
GenBank mRNA JI863066.1 - 6 - 7 836 96

View Full Product Details