Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C4bpb
Entrez Gene ID
100773000
Gene Name
complement component 4 binding protein, beta
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613661: 2730670 - 2740115 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007616518.1 XP_007614708.1 2 - 3 452 96

View Full Product Details