Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tusc5
Entrez Gene ID
100757040
Gene Name
tumor suppressor candidate 5
Gene Aliases
I79_013354
Chromosome Location
Chr.Un|NW_003614210: 161963 - 178892 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506090.1 XP_003506138.1 2 - 3 512 64

View Full Product Details