Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC100759463
Entrez Gene ID
100759463
Gene Name
chromosome unknown open reading frame, human C11orf85
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614946: 332838 - 353377 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510717.1 XP_003510765.1 7 - 8 536 64

View Full Product Details