Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcyox1l
Entrez Gene ID
100758588
Gene Name
prenylcysteine oxidase 1 like
Gene Aliases
I79_016589
Chromosome Location
Chr.Un|NW_003614565: 248293 - 255587 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649241.1 XP_007647431.1 2 - 3 487 67

View Full Product Details