Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcns1
Entrez Gene ID
100762339
Gene Name
potassium voltage-gated channel, delayed-rectifier, subfamily S, member 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614077: 540395 - 544883 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504588.1 XP_003504636.1 1 - 2 1028 74

View Full Product Details