Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zglp1
Entrez Gene ID
100761549
Gene Name
zinc finger, GATA-like protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613752: 1164018 - 1168503 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499742.2 XP_003499790.1 1 - 2 518 65

View Full Product Details