Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zic4
Entrez Gene ID
100755513
Gene Name
Zic family member 4
Gene Aliases
I79_000874
Chromosome Location
Chr.Un|NW_003613597: 331763 - 337216 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495616.2 XP_003495664.1 1 - 2 706 92

View Full Product Details