Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nccrp1
Entrez Gene ID
100770916
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Gene Aliases
I79_010770
Chromosome Location
Chr.Un|NW_003614005: 1063661 - 1088354 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644416.1 XP_007642606.1 5 - 6 688 72

View Full Product Details