Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab33a
Entrez Gene ID
100771082
Gene Name
RAB33A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613712: 200094 - 211988 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498975.2 XP_003499023.1 1 - 2 257 69

View Full Product Details