Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccl1
Entrez Gene ID
103159565
Gene Name
chemokine (C-C motif) ligand 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613601: 3830532 - 3833307 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644150.1 XP_007642340.1 1 - 2 78 73

View Full Product Details