Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vsig10l
Entrez Gene ID
100754269
Gene Name
V-set and immunoglobulin domain containing 10 like
Gene Aliases
I79_021696
Chromosome Location
Chr.Un|NW_003615616: 151434 - 158637 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653251.1 XP_007651441.1 7 - 8 2278 78

View Full Product Details