Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ca13
Entrez Gene ID
100771949
Gene Name
carbonic anhydrase XIII
Gene Aliases
I79_004472
Chromosome Location
Chr.Un|NW_003613698: 2150499 - 2173758 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007630407.1 XP_007628597.1 5 - 6 679 103

View Full Product Details