Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nrip2
Entrez Gene ID
100769051
Gene Name
nuclear receptor interacting protein 2
Gene Aliases
I79_001347
Chromosome Location
Chr.Un|NW_003613610: 313850 - 322804 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496017.2 XP_003496065.1 4 - 5 687 67

View Full Product Details