Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo46
Entrez Gene ID
103159278
Gene Name
F-box protein 46
Gene Aliases
I79_012665
Chromosome Location
Chr.Un|NW_003614150: 811395 - 814166 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645977.1 XP_007644167.1 2 - 3 868 112

View Full Product Details