Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nudt15
Entrez Gene ID
100761298
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 15
Gene Aliases
I79_016344
Chromosome Location
Chr.Un|NW_003614537: 27757 - 33055 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508407.2 XP_003508455.1 2 - 3 384 78

View Full Product Details