Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Esf1
Entrez Gene ID
100771608
Gene Name
ESF1, nucleolar pre-rRNA processing protein, homolog (S. cerevisiae)
Gene Aliases
ESF1-like, I79_015571
Chromosome Location
Chr.Un|NW_003614054: 388463 - 408698 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644875.1 XP_007643065.1 4 - 5 453 106
GenBank mRNA JI868725.1 - 4 - 5 399 106

View Full Product Details