Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp4r2
Entrez Gene ID
100758228
Gene Name
protein phosphatase 4, regulatory subunit 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613585: 1359387 - 1402550 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007620356.1 XP_007618546.1 1 - 2 68 102

View Full Product Details