Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem8c
Entrez Gene ID
100752392
Gene Name
transmembrane protein 8C
Gene Aliases
I79_025167
Chromosome Location
Chr.Un|NW_003618467: 8272 - 16979 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515560.2 XP_003515608.1 2 - 3 246 105

View Full Product Details