Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hes1
Entrez Gene ID
100752756
Gene Name
hes family bHLH transcription factor 1
Gene Aliases
I79_015503
Chromosome Location
Chr.Un|NW_003614422: 734616 - 736283 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648368.1 XP_007646558.1 2 - 3 469 80
GenBank mRNA JI894162.1 - 2 - 3 473 80

View Full Product Details