Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fibcd1
Entrez Gene ID
100767519
Gene Name
fibrinogen C domain containing 1
Gene Aliases
I79_022764
Chromosome Location
Chr.Un|NW_003616105: 7684 - 35011 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513748.1 XP_003513796.1 2 - 3 556 61

View Full Product Details