Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr16
Entrez Gene ID
100774540
Gene Name
WD repeat domain 16
Gene Aliases
I79_002689
Chromosome Location
Chr.Un|NW_003613643: 13456 - 50527 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007608156.1 XP_007606346.1 4 - 5 445 97

View Full Product Details