Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tshz2
Entrez Gene ID
100759966
Gene Name
teashirt zinc finger homeobox 2
Gene Aliases
I79_024603
Chromosome Location
Chr.Un|NW_003617456: 14190 - 17497 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007655155.1 XP_007653345.1 1 - 1 1620 83

View Full Product Details