Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tatdn2
Entrez Gene ID
100752200
Gene Name
TatD DNase domain containing 2
Gene Aliases
I79_002354
Chromosome Location
Chr.Un|NW_003613637: 751896 - 768087 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653856.1 XP_007652046.1 7 - 8 2624 82

View Full Product Details