Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sncb
Entrez Gene ID
100774653
Gene Name
synuclein, beta
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613681: 1479506 - 1485637 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498163.1 XP_003498211.1 2 - 3 163 59

View Full Product Details