Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sirpb1
Entrez Gene ID
100751767
Gene Name
signal-regulatory protein beta 1
Gene Aliases
I79_017160
Chromosome Location
Chr.Un|NW_003614639: 477663 - 502927 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509067.2 XP_003509115.1 4 - 5 1013 100

View Full Product Details