Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc39a2
Entrez Gene ID
100752281
Gene Name
solute carrier family 39 (zinc transporter), member 2
Gene Aliases
I79_023883
Chromosome Location
Chr.Un|NW_003616775: 79444 - 81166 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514617.2 XP_003514665.1 2 - 3 246 106

View Full Product Details