Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl21c
Entrez Gene ID
100764605
Gene Name
methyltransferase like 21C
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615687: 129253 - 134811 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653381.1 XP_007651571.1 1 - 2 159 83

View Full Product Details