Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hrc
Entrez Gene ID
100771220
Gene Name
histidine rich calcium binding protein
Gene Aliases
I79_016655
Chromosome Location
Chr.Un|NW_003614570: 404671 - 408045 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508642.2 XP_003508690.1 2 - 3 1991 73

View Full Product Details