Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc4b
Entrez Gene ID
100758215
Gene Name
leucine rich repeat containing 4B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614844: 396023 - 414532 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510210.1 XP_003510258.1 1 - 2 301 63

View Full Product Details