Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lipt2
Entrez Gene ID
100756678
Gene Name
lipoyl(octanoyl) transferase 2 (putative)
Gene Aliases
I79_020280
Chromosome Location
Chr.Un|NW_003615205: 35758 - 37392 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511692.2 XP_003511740.1 1 - 2 520 57

View Full Product Details