Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spin4
Entrez Gene ID
100767892
Gene Name
spindlin family, member 4
Gene Aliases
Spindlin-4
Chromosome Location
Chr.Un|NW_003615217: 74788 - 84913 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652248.1 XP_007650438.1 1 - 1 1113 95
XM_007652249.1 XP_007650439.1 1 - 1 1113 95

View Full Product Details