Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nptxr
Entrez Gene ID
100750799
Gene Name
neuronal pentraxin receptor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614714: 373646 - 382491 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650060.1 XP_007648250.1 2 - 3 274 96

View Full Product Details