Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc57
Entrez Gene ID
100771842
Gene Name
coiled-coil domain containing 57
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613637: 3066508 - 3169748 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654255.1 XP_007652445.1 14 - 15 2299 96
XM_007654258.1 XP_007652448.1 13 - 14 2174 96
XM_007654261.1 XP_007652451.1 13 - 14 2141 96
GenBank mRNA JI873672.1 - 2 - 3 250 96

View Full Product Details