Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdf
Entrez Gene ID
100762250
Gene Name
peptide deformylase (mitochondrial)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614572: 23765 - 26868 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508692.2 XP_003508740.1 1 - 2 160 63

View Full Product Details