Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dok3
Entrez Gene ID
100763567
Gene Name
docking protein 3
Gene Aliases
I79_003891
Chromosome Location
Chr.Un|NW_003613681: 749529 - 755132 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007623190.1 XP_007621380.1 4 - 5 1506 64

View Full Product Details