Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt3a
Entrez Gene ID
100764164
Gene Name
wingless-type MMTV integration site family, member 3A
Gene Aliases
I79_010136
Chromosome Location
Chr.Un|NW_003613960: 488942 - 522897 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643947.1 XP_007642137.1 4 - 5 289 67

View Full Product Details