Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpl34
Entrez Gene ID
100753386
Gene Name
ribosomal protein L34
Gene Aliases
I79_023163
Chromosome Location
Chr.Un|NW_003616312: 106509 - 109195 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514042.2 XP_003514090.1 2 - 3 206 87
GenBank mRNA JI882894.1 - - 87

View Full Product Details